missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylphenoxy)aniline, 96%, Thermo Scientific™

Product Code. 15456997
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456997 10 g 10g
15446997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456997 Supplier Thermo Scientific Alfa Aesar Supplier No. H34084.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56705-83-0
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.25
MDL Number MFCD02663364
InChI Key JPCCVWJJMUIBJR-UHFFFAOYSA-N
PubChem CID 92505
IUPAC Name 4-(2-methylphenoxy)aniline
SMILES CC1=CC=CC=C1OC1=CC=C(N)C=C1

Specifications

Melting Point 61°C to 62°C
Quantity 10 g
Synonym 4-2-methylphenoxy aniline, 4-o-tolyloxy aniline, 4-2-methylphenoxy phenyl amine, 4-2-methylphenoxy phenylamine, benzenamine,4-2-methylphenoxy, benzenamine, 4-2-methylphenoxy, 4-o-tolyloxyphenylamine, 4-2-methylphenoxy benzenamine
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material 4-(2-Methylphenoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.