Learn More
4-(2-Methylphenyl)-3-thiosemicarbazide, 98%, Thermo Scientific™
Artikelnummer. 11347396
Ansicht ändern
Click to view available options
Quantity:
5 g
25 g
Packungsgröße:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347396 5 g 5g
11357396 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11347396 Supplier Thermo Scientific Alfa Aesar Supplier No. L09382.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 614-10-8
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00041277
InChI Key QCMQZMITIOLXFZ-UHFFFAOYSA-N
Synonym 4-2-methylphenyl-3-thiosemicarbazide, n-o-tolyl hydrazinecarbothioamide, 3-amino-1-2-methylphenyl thiourea, n-2-methylphenyl hydrazinecarbothioamide, hydrazino 2-methylphenyl amino methane-1-thione, acmc-1bcwm, wln: zmyus&mr b1, 4-o-tolyl thiosemicarbazide, 1-amino-3-2-methylphenyl thiourea, hydrazinecarbothioamide,n-2-methylphenyl
PubChem CID 737133
IUPAC Name 1-amino-3-(2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(=S)NN
Specifications
Melting Point 145°C to 148°C
Quantity 5 g
UN Number UN2811
Beilstein 880437
Formula Weight 181.26
Percent Purity 98%
Chemical Name or Material 4-(2-Methylphenyl)-3-thiosemicarbazide