missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridinyl)benzoyl Chloride Hydrochloride, TRC

Product Code. 30417961
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417961 2.5 g 2.5g
30415908 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417961 Supplier Toronto Research Chemicals Supplier No. TRCP6758902.5G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223128-24-3
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 254.11
Synonym 4-(Pyridin-2-yl)benzoyl Chloride
IUPAC Name 4-pyridin-2-ylbenzoyl chloride;hydrochloride
SMILES Cl.ClC(=O)c1ccc(cc1)c2ccccn2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H8ClNO.ClH/c13-12(15)10-6-4-9(5-7-10)11-3-1-2-8-14-11;/h1-8H;1H
Formula Weight 253.0061
Chemical Name or Material 4-(2-Pyridinyl)benzoyl Chloride Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.