missing translation for 'onlineSavingsMsg'
Learn More

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 11386855
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386855 5 g Amber Glass Bottle 5g
10183454 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386855 Supplier Thermo Scientific Maybridge Supplier No. CC68216DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56935-79-6
Molecular Formula C12H11N3
Molecular Weight (g/mol) 197.241
MDL Number MFCD09817559
InChI Key BTXLPDNFEZIQBZ-UHFFFAOYSA-N
Synonym 4-3,5-dimethyl-1h-pyrazol-1-yl benzonitrile, 4-3,5-dimethylpyrazol-1-yl benzonitrile, acmc-209luz, 4-3,5-dimethylpyrazol-1-yl-benzonitrile, 4-3,5-dimethylpyrazolyl benzenecarbonitrile, 1-4-cyanophenyl-3,5-dimethyl-1h-pyrazole, benzonitrile, 4-3,5-dimethyl-1h-pyrazol-1-yl
PubChem CID 15005387
IUPAC Name 4-(3,5-dimethylpyrazol-1-yl)benzonitrile
SMILES CC1=CC(=NN1C2=CC=C(C=C2)C#N)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 197.24
Percent Purity 97%
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.