missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine, 97%

Product Code. 11439560
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439560 250 mg 250mg
11449560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439560 Supplier Thermo Scientific Alfa Aesar Supplier No. H50873.MD
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 733748-92-0 | C10H17N3O | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 733748-92-0
Molecular Formula C10H17N3O
Molecular Weight (g/mol) 195.27
MDL Number MFCD08061038
InChI Key QDRJZNBKSMAEIE-UHFFFAOYSA-N
Synonym 4-3-isopropyl-1,2,4-oxadiazol-5-yl piperidine, 3-isopropyl-5-piperidin-4-yl-1,2,4-oxadiazole, 4-3-isopropyl-1,2,4 oxadiazol-5-yl-piperidine, 5-piperidin-4-yl-3-propan-2-yl-1,2,4-oxadiazole, piperidine, 4-3-1-methylethyl-1,2,4-oxadiazol-5-yl, 4-3-propan-2-yl-1,2,4-oxadiazol-5-yl piperidine, 4-3-1-methylethyl-1,2,4-oxadiazol-5-yl piperidine, 3-isopropyl-5-4-piperidyl-1,2,4-oxadiazole, 3-methylethyl-5-4-piperidyl-1,2,4-oxadiazole
PubChem CID 24208845
IUPAC Name 5-piperidin-4-yl-3-propan-2-yl-1,2,4-oxadiazole
SMILES CC(C)C1=NOC(=N1)C1CCNCC1

Specifications

Refractive Index 1.486
Quantity 250 mg
Formula Weight 195.27
Percent Purity 97%
Chemical Name or Material 4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.