missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol Hydrochloride (Duloxetine Impurity), TRC

Product Code. 30011144
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011144 10 mg 10mg
30007936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011144 Supplier Toronto Research Chemicals Supplier No. TRCM28720010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953028-76-7
Molecular Formula C18H19NOS.ClH
Molecular Weight (g/mol) 333.88
Synonym Duloxetine Impurity
IUPAC Name 4-[3-(methylamino)-1-(2-thienyl)propyl]naphthalen-1-ol;hydrochloride
SMILES Cl.CNCCC(c1cccs1)c2ccc(O)c3ccccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H19NOS.ClH/c1-19-11-10-16(18-7-4-12-21-18)14-8-9-17(20)15-6-3-2-5-13(14)15;/h2-9,12,16,19-20H,10-11H2,1H3;1H
Formula Weight 333.0954
Chemical Name or Material 4-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol Hydrochloride(Duloxetine Impurity)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.