missing translation for 'onlineSavingsMsg'
Learn More

[[4-[(3-Methylphenyl)amino]-3-pyridinyl]sulfonyl]carbamic Acid Ethyl Ester, TRC

Product Code. 30137029
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137029 1 g 1g
30009038 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137029 Supplier Toronto Research Chemicals Supplier No. TRCM3206851G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72810-57-2
Molecular Formula C15 H17 N3 O4 S
Molecular Weight (g/mol) 335.38
Synonym Ethyl [[4-[(3-Methylphenyl)amino]pyridin-3-yl]sulfonyl]carbamate, Torasemide, Anhydrous Imp. E (EP)
IUPAC Name ethyl N-[4-(3-methylanilino)pyridin-3-yl]sulfonylcarbamate
SMILES CCOC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H17N3O4S/c1-3-22-15(19)18-23(20,21)14-10-16-8-7-13(14)17-12-6-4-5-11(2)9-12/h4-10H,3H2,1-2H3,(H,16,17)(H,18,19)
Formula Weight 335.094
Chemical Name or Material ethyl [(4-[(3-methylphenyl)amino]pyridin-3-yl)sulfonyl]carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.