Learn More
4-(3-Oxopiperazin-1-yl)benzonitrile, TRC
Product Code. 30415493
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415493 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30415493 Supplier Toronto Research Chemicals Supplier No. TRCO990245250MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(3-Oxopiperazin-1-yl)benzonitrile
CAS 1019601-14-9
Molecular Formula C11H11N3O
Quantity 250 mg
InChI Formula InChI=1S/C11H11N3O/c12-7-9-1-3-10(4-2-9)14-6-5-13-11(15)8-14/h1-4H,5-6,8H2,(H,13,15)
SMILES O=C1CN(CCN1)c2ccc(cc2)C#N
IUPAC Name 4-(3-oxopiperazin-1-yl)benzonitrile
Molecular Weight (g/mol) 201.22
Formula Weight 201.0902