missing translation for 'onlineSavingsMsg'
Learn More

4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine, 97%, Thermo Scientific™

Product Code. 10327154
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327154 1 g Amber Glass Bottle 1g
10422264 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10327154 Supplier Thermo Scientific Maybridge Supplier No. CC74239DA
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 364794-85-4
Molecular Formula C15H24BNO3S
Molecular Weight (g/mol) 309.23
MDL Number MFCD11841079
InChI Key AMUMGAXTCHBNPU-UHFFFAOYSA-N
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
PubChem CID 23438311
IUPAC Name 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 309.24
Percent Purity 97%
Chemical Name or Material 4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.