Learn More
4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane), 98%, BLD Pharm™
Specifications
Specifications
| CAS | 230299-21-5 |
| Molecular Formula | C12H24B2O4 |
| MDL Number | MFCD06246008 |
| Quantity | 500 g |
| SMILES | CC1CC(C)(C)OB(B2OC(C)CC(C)(C)O2)O1 |
| Molecular Weight (g/mol) | 253.94 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane) |