missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 98%, BLD Pharm™

Product Code. 30797922
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30797922 500 g 500g
30790942 5 g 5g
30789918 10 g 10g
30795497 25 g 25g
30796277 1 g 1g
30794755 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30797922 Supplier BLD Pharm Supplier No. BD14831500g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 180516-87-4
Molecular Formula C13H17BO4
MDL Number MFCD01863710
Quantity 500 g
SMILES C1=C(C=CC(=C1)C(=O)O)B2OC(C(O2)(C)C)(C)C
Molecular Weight (g/mol) 248.08
Percent Purity 98%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.