missing translation for 'onlineSavingsMsg'
Learn More

4,4',4' '-Methylidynetrisbenzonitrile (Letrozole Impurity), TRC

Product Code. 17932495
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932495 10 mg 10mg
17923425 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17932495 Supplier Toronto Research Chemicals Supplier No. TRCM31295010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113402-31-6
Molecular Formula C22 H13 N3
Molecular Weight (g/mol) 319.36
Synonym Letrozole Impurity B
IUPAC Name 4-[bis(4-cyanophenyl)methyl]benzonitrile
SMILES N#Cc1ccc(cc1)C(c2ccc(cc2)C#N)c3ccc(cc3)C#N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H13N3/c23-13-16-1-7-19(8-2-16)22(20-9-3-17(14-24)4-10-20)21-11-5-18(15-25)6-12-21/h1-12,22H
Formula Weight 319.1109
Percent Purity >95%
Chemical Name or Material 4,4',4' '-Methylidynetrisbenzonitrile
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.