missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-oxobutanoic Acid Ethyl Ester, TRC

Product Code. 30454276
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454276 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454276 Supplier Toronto Research Chemicals Supplier No. TRCT7920602.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4,4-Trifluoro-3-oxobutanoic Acid Ethyl Ester
CAS 372-31-6
Molecular Formula C6H7F3O3
Quantity 2.5 g
Synonym 4,4,4-Trifluoroacetoacetic Acid Ethyl Ester, 1-Ethoxy-4,4,4-trifluorobutane-1,3-dione, 4,4,4-Trifluoro-3-oxo-butyric Acid Ethyl Ester, 4,4,4-Trifluoro-3-oxobutanoic Acid Ethyl Ester, 4,4,4-Trifluoroacetoacetic Acid Ethyl Ester, Ethyl (Trifluoroacetyl)acetate, Ethyl 3-oxo-4,4,4-Trifluorobutanoate, Ethyl 4,4,4-Trifluoro-3-oxobutanoate, Ethyl 4,4,4-Trifluoro-3-oxobutyrate, Ethyl 4,4,4-Trifluoroacetoacetate, Ethyl 4,4,4-Trifluoroacetylacetonate, Ethyl Trifluoroacetoacetate, Ethyl gamma,gamma,gamma-Trifluoroacetoacetate, Ethyl ω,ω,ω-Trifluoroacetoacetate, NSC 42739, NSC 49750
InChI Formula InChI=1S/C6H7F3O3/c1-2-12-5(11)3-4(10)6(7,8)9/h2-3H2,1H3
SMILES CCOC(=O)CC(=O)C(F)(F)F
IUPAC Name ethyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol) 184.11
Formula Weight 184.0347
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.