missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-(trifluoromethyl)butanoic Acid, TRC

Product Code. 30453599
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453599 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453599 Supplier Toronto Research Chemicals Supplier No. TRCT791495250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4,4-Trifluoro-3-(trifluoromethyl)butanoic Acid
CAS 17327-33-2
Molecular Formula C5H4F6O2
Quantity 250 mg
Synonym 4,4,4-Trifluoro-3-trifluoromethylbutyric Acid, 3-Trifluoromethyl-4,4,4-trifluorobutanoic Acid
InChI Formula InChI=1S/C5H4F6O2/c6-4(7,8)2(1-3(12)13)5(9,10)11/h2H,1H2,(H,12,13)
SMILES OC[C@@H](O)C=O
Molecular Weight (g/mol) 210.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.