missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 15478305
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478305 250 mg 250mg
15468305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478305 Supplier Thermo Scientific Alfa Aesar Supplier No. H33086.MD
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1174064-62-0
Molecular Formula C9H6BrClN2O2S
Molecular Weight (g/mol) 321.573
MDL Number MFCD12406835
InChI Key LRYXAJMNOSSJDD-UHFFFAOYSA-N
Synonym 4-4-bromo-1h-pyrazol-1-yl benzenesulfonyl chloride, 4-4-bromopyrazol-1-yl benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl-benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl benzene-1-sulfonyl chloride
PubChem CID 45594324
IUPAC Name 4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1N2C=C(C=N2)Br)S(=O)(=O)Cl

Specifications

Melting Point 144°C to 146°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 321.58
Percent Purity 95%
Chemical Name or Material 4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.