missing translation for 'onlineSavingsMsg'
Learn More

4,4'-cis-Stilbenedicarboxylic acid, 95%

Product Code. 11386906
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386906 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11386906 Supplier Thermo Scientific Alfa Aesar Supplier No. L04364.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 133005-88-6 | C16H12O4 | 268.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 133005-88-6
Melting Point >300°C
Molecular Formula C16H12O4
MDL Number MFCD00013994
Quantity 1 g
Beilstein 1978660
Synonym 4-z-2-4-carboxyphenyl ethenyl benzoic acid, cis-stilbene-4,4'-dicarboxylic acid, z-4,4'-ethene-1,2-diyl dibenzoic acid, cis-4,4'-dicarboxystilbene, z-4,4'-stilbene dicarboxylic acid, z-stilbene-4,4'-dicarboxylic acid, 4,4'-cis-stilbenedicarboxylic acid
InChI Key SBBQDUFLZGOASY-UPHRSURJSA-N
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)C(=O)O)C(=O)O
IUPAC Name 4-[(Z)-2-(4-carboxyphenyl)ethenyl]benzoic acid
Molecular Weight (g/mol) 268.268
PubChem CID 18647961
Formula Weight 268.27
Percent Purity 95%
Chemical Name or Material 4,4'-cis-Stilbenedicarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.