missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbiphenyl, 97%, Thermo Scientific™

Product Code. 10356370 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356370 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10356370 Supplier Thermo Scientific Supplier No. 116080010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 613-33-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 117°C to 122°C
Color White to Yellow
Boiling Point 295°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C14H14
Linear Formula CH3C6H4C6H4CH3
MDL Number MFCD00008545
Quantity 1 g
Beilstein 05, 610
Synonym 4,4'-dimethylbiphenyl, 4,4'-bitolyl, 4,4'-dimethyldiphenyl, bi-p-tolyl, di-p-tolyl, p,p'-bitolyl, 4,4'-dimethyl-1,1'-biphenyl, 4,4'-ditolyl, 1,1'-biphenyl, 4,4'-dimethyl, 4,4'-dimethyl-biphenyl
InChI Key RZTDESRVPFKCBH-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C
IUPAC Name 1-methyl-4-(4-methylphenyl)benzene
Molecular Weight (g/mol) 182.27
PubChem CID 11941
Formula Weight 182.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbiphenyl, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.