missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dinitrocarbanilide-d8 (rings-d8), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis-(4-nitrophenyl)urea D8 |
| CAS | 1156508-87-0 |
| Molecular Formula | C13 2H8 H2 N4 O5 |
| Quantity | 0.05 g |
| Synonym | 1,3-Bis(4-nitrophenyl)urea-d8, N,N'-Bis-(4-nitrophenyl)urea D8, Urea, N,N'-bis(4-nitrophenyl-2,3,5,6-d4)-, N,N'-Bis(4-nitrophenyl-2,3,5,6-d4)urea |
| InChI Formula | InChI=1 S/C13H10N4O5/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22/h1-8 H,(H2,14,15,18)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1NC(=O)Nc2c([2 H])c([2 H])c(c([2 H])c2[2 H])[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 1,3-bis(2,3,5,6-tetradeuterio-4-nitrophenyl)urea |
| Molecular Weight (g/mol) | 310.29 |
| Formula Weight | 310.1153 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction