missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxy-3-methylbenzenesulfonyl)-2-methylphenol, TRC

Product Code. 30400370
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400370 50 mg
30389532 25 mg
30392791 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400370 Supplier Toronto Research Chemicals Supplier No. TRCH94774350MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16346-97-7
Molecular Formula C14 H14 O4 S
Molecular Weight (g/mol) 278.32
Synonym 4,4′-Sulfonylbis(2-methylphenol)
IUPAC Name 4-(4-hydroxy-3-methyl-phenyl)sulfonyl-2-methyl-phenol
SMILES Cc1cc(ccc1O)S(=O)(=O)c2ccc(O)c(C)c2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H14O4S/c1-9-7-11(3-5-13(9)15)19(17,18)12-4-6-14(16)10(2)8-12/h3-8,15-16H,1-2H3
Formula Weight 278.0613
Chemical Name or Material 4-(4-Hydroxy-3-methylbenzenesulfonyl)-2-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.