missing translation for 'onlineSavingsMsg'
Learn More

4,4'-[(Phenylmethylene)bis[(2,5-dimethyl-4,1-phenylene)-2,1-diazenediyl]]bis[3-hydroxy-N-phenyl-2-naphthalenecarboxamide, TRC

Product Code. 30424399
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424399 500 mg
30425000 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424399 Supplier Toronto Research Chemicals Supplier No. TRCP335910500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60033-00-3
Molecular Formula C57H46N6O4
Molecular Weight (g/mol) 879.01
IUPAC Name 3-hydroxy-4-[(E)-[4-[[4-[(E)-[2-hydroxy-3-(phenylcarbamoyl)naphthalen-1-yl]diazenyl]-2,5-dimethylphenyl]-phenylmethyl]-2,5-dimethylphenyl]diazenyl]-N-phenylnaphthalene-2-carboxamide
SMILES Cc1cc(C(c2ccccc2)c3cc(C)c(cc3C)N=Nc4c(O)c(cc5ccccc45)C(=O)Nc6ccccc6)c(C)cc1N=Nc7c(O)c(cc8ccccc78)C(=O)Nc9ccccc9

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C57H46N6O4/c1-34-30-49(60-62-52-43-26-16-14-20-39(43)32-47(54(52)64)56(66)58-41-22-10-6-11-23-41)36(3)28-45(34)51(38-18-8-5-9-19-38)46-29-37(4)50(31-35(46)2)61-63-53-44-27-17-15-21-40(44)33-48(55(53)65)57(67)59-42-24-12-7-13-25-42/h5-33,51,64-65H,1-4H3,(H,58,66)(H,59,67)
Formula Weight 878.3581
Chemical Name or Material 4,4'-[(Phenylmethylene)bis((2,5-dimethyl-4,1-phenylene)-2,1-diazenediyl)]bis-3-hydroxy-N-phenyl-2-naphthalenecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.