Learn More
4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 3543-75-7 |
| Molecular Formula | C16H22Cl3N3O2 |
| MDL Number | MFCD01658758 |
| Quantity | 100 mg |
| SMILES | Cl.CN1C(CCCC(O)=O)=NC2=C1C=CC(=C2)N(CCCl)CCCl |
| Molecular Weight (g/mol) | 394.72 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride |