missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide, TRC

Product Code. 30012197
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012197 10 mg 10mg
30009045 100 mg 100mg
17983105 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012197 Supplier Toronto Research Chemicals Supplier No. TRCD43558810MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1770812-37-7
Molecular Formula C19 H17 Cl2 N3 O5 S
Molecular Weight (g/mol) 470.33
Synonym 2-Thiophenecarboxamide, 4,5-dichloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-, 4,5-Dichloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide, (S)-4,5-Dichloro-N-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)thiophene-2-carboxamide
IUPAC Name 4,5-dichloro-N-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES Clc1cc(sc1Cl)C(=O)NC[C@H]2CN(C(=O)O2)c3ccc(cc3)N4CCOCC4=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H17Cl2N3O5S/c20-14-7-15(30-17(14)21)18(26)22-8-13-9-24(19(27)29-13)12-3-1-11(2-4-12)23-5-6-28-10-16(23)25/h1-4,7,13H,5-6,8-10H2,(H,22,26)/t13-/m0/s1
Formula Weight 469.0266
Percent Purity >95%
Chemical Name or Material 4,5-Dichloro-N-([(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl)-2-thiophenecarboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.