missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid |
| CAS | 72909-34-3 |
| Molecular Formula | C14 H6 N2 O8 |
| Quantity | 1 mg |
| Synonym | 1H-Pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 4,5-dihydro-4,5-dioxo-, 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 2,7,9-Tricarboxy-4,5-dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline, Coenzyme PQQ, Methoxatin, Methoxatine, PQQ, Pyrroloquinoline quinone |
| InChI Formula | InChI=1S/C14H6N2O8/c17-10-4-2-6(14(23)24)15-8(4)7-3(12(19)20)1-5(13(21)22)16-9(7)11(10)18/h1-2,15H,(H,19,20)(H,21,22)(H,23,24) |
| SMILES | OC(=O)c1cc(C(=O)O)c2c(n1)C(=O)C(=O)c3cc([nH]c23)C(=O)O |
| IUPAC Name | 4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid |
| Molecular Weight (g/mol) | 330.21 |
| Formula Weight | 330.0124 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?