missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminopyrimidine Hemisulfate, Monohydrate 98%, Thermo Scientific™

Product Code. 10570371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10570371 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10570371 Supplier Thermo Scientific Acros Supplier No. 112410050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 207226-28-6, 77709-02-5
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C8H16N8O5S
MDL Number MFCD00149655
Quantity 5g
Synonym 4,6-diaminopyrimidine hemisulfate salt monohydrate, sulfuric acid-pyrimidine-4,6-diamine-water 1/2/1, bis pyrimidine-4,6-diamine sulfuric acid hydrate
InChI Key ZMRLIHKEUKNTRB-UHFFFAOYSA-N
SMILES C1=C(N=CN=C1N)N.C1=C(N=CN=C1N)N.O.OS(=O)(=O)O
IUPAC Name pyrimidine-4,6-diamine;sulfuric acid;hydrate
Molecular Weight (g/mol) 336.327
PubChem CID 54772454
Formula Weight 177.18
Percent Purity 98%
Chemical Name or Material 4,6-Diaminopyrimidine hemisulfate monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.