Learn More
4,6-Dichloro-5-nitropyrimidine, 98%
Product Code. 15432708
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15432708 25 g 25g
15412708 1 g 1g
15422708 5 g 5g
3 options
Product Code. 15432708
Brand: Thermo Scientific Alfa Aesar H56252.14
Request Bulk or Custom Format
CAS: 4316-93-2 | C4HCl2N3O2 | 193.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4316-93-2
Molecular Formula C4HCl2N3O2
Molecular Weight (g/mol) 193.97
MDL Number MFCD00006107
InChI Key HCTISZQLTGAYOX-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro-5-nitro, 4,6-dichloro-5-nitro-pyrimidine, 4,6-dichloro-5-nitro-pyridine, pubchem7078, acmc-1an8c, 4-23-00-00899 beilstein handbook reference, ksc495i5h, 4,6-dichloro-5-nitroprimidine, 4,6 dichloro-5-nitropyrimidine, 5-nitro 4,6-dichloropyrimidine
PubChem CID 20312
IUPAC Name 4,6-dichloro-5-nitropyrimidine
SMILES [O-][N+](=O)C1=C(Cl)N=CN=C1Cl
Specifications
Melting Point 100°C to 103°C
Quantity 25 g
Beilstein 162029
Formula Weight 193.98
Percent Purity 98%
Chemical Name or Material 4,6-Dichloro-5-nitropyrimidine