accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4316-93-2

ONOClNNCl

CAS RN 4316-93-2

IUPAC Name: 4,6-dichloro-5-nitropyrimidine
Synonyms: 4,6-dichloro-5-nitropyrimidine
Molecular Weight (g/mol): 193.97
Molecular Formula: C4HCl2N3O2
InChi Key: HCTISZQLTGAYOX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)N=CN=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)