missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine, TRC

Product Code. 30373304
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373304 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373304 Supplier Toronto Research Chemicals Supplier No. TRCD43583550G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Dichloropyrimidine
CAS 1193-21-1
Molecular Formula C4 H2 Cl2 N2
Quantity 50 g
InChI Formula InChI=1S/C4H2Cl2N2/c5-3-1-4(6)8-2-7-3/h1-2H
SMILES Clc1cc(Cl)ncn1
IUPAC Name 4,6-dichloropyrimidine
Molecular Weight (g/mol) 148.98
Formula Weight 147.9595
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.