missing translation for 'onlineSavingsMsg'
Learn More

4-(6-hydrazinylpyrimidin-4-yl)morpholine, TRC

Product Code. 30402279
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402279 50 mg 50mg
30401317 250 mg -
30400787 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402279 Supplier Toronto Research Chemicals Supplier No. TRCH95579850MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5767-36-2
Molecular Formula C8 H13 N5 O
Molecular Weight (g/mol) 195.222
IUPAC Name (6-morpholin-4-ylpyrimidin-4-yl)hydrazine
SMILES NNc1cc(ncn1)N2CCOCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H13N5O/c9-12-7-5-8(11-6-10-7)13-1-3-14-4-2-13/h5-6H,1-4,9H2,(H,10,11,12)
Formula Weight 195.112
Chemical Name or Material 4-(6-hydrazinylpyrimidin-4-yl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.