Learn More
4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile, TRC
Product Code. 30374246
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374246 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30374246 Supplier Toronto Research Chemicals Supplier No. TRCD45347510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
CAS 1019853-64-5
Molecular Formula C26H19N5O
Quantity 10 mg
InChI Formula InChI=1S/C26H19N5O/c27-16-18-17-28-23-22(18)24(32)30-25(29-23)31-26(19-10-4-1-5-11-19,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,17H,(H3,28,29,30,31,32)
SMILES O=C1C2=C(NC=C2C#N)N=C(NC(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)N1
IUPAC Name 4-oxo-2-(tritylamino)-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
Molecular Weight (g/mol) 417.46
Formula Weight 417.16