missing translation for 'onlineSavingsMsg'
Learn More

4,8,10-Trimethoxy Urolithin E, TRC

Product Code. 30457046
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457046 50 mg 50mg
30453434 25 mg 25mg
30455439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457046 Supplier Toronto Research Chemicals Supplier No. TRCT84703550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,8,10-Trimethoxy Urolithin E
Analyte or Component Names 4,8,10-Trimethoxy Urolithin E
Shipping Condition Room Temperature
Molecular Formula C16 H14 O6
Synonym 3-Hydroxy-4,8,10-trimethoxy-6H-benzo[c]chromen-6-one
InChI Formula InChI=1S/C16H14O6/c1-19-8-6-10-13(12(7-8)20-2)9-4-5-11(17)15(21-3)14(9)22-16(10)18/h4-7,17H,1-3H3
Solution Type Neat
SMILES COc1cc(OC)c2c(c1)C(=O)Oc3c(OC)c(O)ccc23
IUPAC Name 3-hydroxy-4,8,10-trimethoxybenzo[c]chromen-6-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 302.279
Quantity 50 mg
Formula Weight 302.079
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.