missing translation for 'onlineSavingsMsg'
Learn More

4-(8-Quinolinylsulfonylamino)benzoic acid, 96%, Thermo Scientific™

Product Code. 15476647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476647 1 g 1g
15438645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476647 Supplier Thermo Scientific Alfa Aesar Supplier No. H33788.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116834-64-1
Molecular Formula C16H12N2O4S
Molecular Weight (g/mol) 328.34
MDL Number MFCD00601229
InChI Key NBQVTLMGAPBCJA-UHFFFAOYSA-N
Synonym 4-quinoline-8-sulfonamido benzoic acid, 4-8-quinolinylsulfonyl amino benzoic acid, benzoicacid, 4-8-quinolinylsulfonyl amino, 4-8-quinolinylsulfonylamino benzoic acid, acmc-20mmuc, cbmicro_030879, cambridge id 5760668, 4-8-quinolylsulfonyl amino benzoic acid, 4-quinolin-8-ylsulfonylamino benzoic acid, 4-quinoline-8-sulfonylamino-benzoic acid
PubChem CID 776645
IUPAC Name 4-(quinolin-8-ylsulfonylamino)benzoic acid
SMILES OC(=O)C1=CC=C(NS(=O)(=O)C2=C3N=CC=CC3=CC=C2)C=C1

Specifications

Melting Point 260°C to 269°C
Quantity 1 g
Formula Weight 328.34
Percent Purity 96%
Chemical Name or Material 4-(8-Quinolinylsulfonylamino)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.