Learn More
4-Acetoxystyrene, TRC
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Acetoxystyrene |
| CAS | 2628-16-2 |
| Molecular Formula | C10H10O2 |
| Quantity | 10 g |
| Synonym | 4-Acetoxystyrene, 4-Ethenylphenol acetate, 4-Vinylphenyl Acetate, PACS, p-Acetoxystyrene |
| InChI Formula | InChI=1S/C10H10O2/c1-3-9-4-6-10(7-5-9)12-8(2)11/h3-7H,1H2,2H3 |
| SMILES | CC(=O)Oc1ccc(C=C)cc1 |
| IUPAC Name | (4-ethenylphenyl) acetate |
| Molecular Weight (g/mol) | 162.19 |
| Formula Weight | 162.0681 |