missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Allyl-2-methoxyphenyl Methanesulfonate, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 4-Allyl-2-methoxyphenyl Methanesulfonate |
| CAS | 91971-63-0 |
| Molecular Formula | C11H14O4S |
| Quantity | 25 mg |
| Synonym | 4-Allyl-2-methoxyphenol Methanesulfonate, 2-Methoxy-4-(2-propen-1-yl)phenol 1-Methanesulfonate, 2-Methoxy-4-(2-propenyl)phenol Methanesulfonate |
| InChI Formula | InChI=1S/C11H14O4S/c1-4-5-9-6-7-10(11(8-9)14-2)15-16(3,12)13/h4,6-8H,1,5H2,2-3H3 |
| SMILES | COC1=C(C=CC(CC=C)=C1)OS(C)(=O)=O |
| IUPAC Name | 4-allyl-2-methoxyphenyl methanesulfonate |
| Molecular Weight (g/mol) | 242.29 |
| Formula Weight | 242.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction