missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, TRC

Product Code. 30309651
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309651 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309651 Supplier Toronto Research Chemicals Supplier No. TRCA5722405G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estragole
CAS 140-67-0
Molecular Formula C10 H12 O
Quantity 5 g
Synonym Anisole, p-allyl- (8CI), Benzene, 1-methoxy-4-(2-propenyl)- (9CI), Estragole (6CI), Ether, p-allylphenyl methyl (7CI), 1-Methoxy-4-(2-propen-1-yl)benzene, 1-Allyl-4-methoxybenzene, 1-Methoxy-4-(2-propenyl)benzene, 3-(4-Methoxyphenyl)-1-propene, 3-(4-Methoxyphenyl)propene, 3-(p-Methoxyphenyl)-1-propene, 3-(p-Methoxyphenyl)propene, 4-Allylanisole, 4-Allylmethoxybenzene, 4-Methoxyallylbenzene, AUSTL 21320, Chavicol methyl ether, Chavicol, O-methyl-, Chavicol, methyl-, Esdragol, Esdragole, Esdragon, Esteragol, Estragenole, Estragol, Isoanethole, Methyl chavicol, NSC 404113, O-Methylchavicol, p-Allylanisole, p-Allylphenyl methyl ether, p-Methoxyallylbenzene
InChI Formula InChI=1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3,5-8H,1,4H2,2H3
SMILES COc1ccc(CC=C)cc1
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
Molecular Weight (g/mol) 148.2
Formula Weight 148.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.