missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-tert-butyl-3-(1’-naphthylmethyl)pyrazolo[3,4-d]pyrimidine, TRC

Product Code. 17933065
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933065 25 mg 25mg
17973255 250 mg 250mg
17950765 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933065 Supplier Toronto Research Chemicals Supplier No. TRCA60300325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221244-14-0
Molecular Formula C20 H21 N5
Molecular Weight (g/mol) 331.41
Synonym 1-NM-PP1
IUPAC Name 1-tert-butyl-3-(naphthalen-1-ylmethyl)pyrazolo[3,4-d]pyrimidin-4-amine
SMILES CC(C)(C)n1nc(Cc2cccc3ccccc23)c4c(N)ncnc14

Specifications

Analyte or Component Names 4-Amino-1-tert-butyl-3-(1'-naphthylmethyl)pyrazolo[3,4-d]pyrimidine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C20H21N5/c1-20(2,3)25-19-17(18(21)22-12-23-19)16(24-25)11-14-9-6-8-13-7-4-5-10-15(13)14/h4-10,12H,11H2,1-3H3,(H2,21,22,23)
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Formula Weight 331.1797
Percent Purity >95%
Chemical Name or Material 4-Amino-1-tert-butyl-3-(1'-naphthylmethyl)pyrazolo[3,4-d]pyrimidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.