missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-mercaptopyrimidine, TRC

Product Code. 30312096
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30312096 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30312096

Brand: Toronto Research Chemicals TRCA61193050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-2-mercaptopyrimidine
CAS 333-49-3
Molecular Formula C4 H5 N3 S
Quantity 50 g
InChI Formula InChI=1S/C4H5N3S/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)
SMILES NC1=NC(=S)NC=C1
IUPAC Name 4-amino-1H-pyrimidine-2-thione
Molecular Weight (g/mol) 127.17
Formula Weight 127.0204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.