missing translation for 'onlineSavingsMsg'
Learn More

(4-Amino-2-methylpyrimidin-5-yl)methyl Trihydrogen Diphosphate, TRC

Product Code. 30138157
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138157 5 mg 5mg
30136433 10 mg 10mg
30012319 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138157 Supplier Toronto Research Chemicals Supplier No. TRCA5782695MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 841-01-0
Molecular Formula C6H11N3O7P2
Molecular Weight (g/mol) 299.11
IUPAC Name (4-amino-2-methylpyrimidin-5-yl)methyl phosphono hydrogen phosphate
SMILES Cc1ncc(COP(=O)(O)OP(=O)(O)O)c(N)n1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H11N3O7P2/c1-4-8-2-5(6(7)9-4)3-15-18(13,14)16-17(10,11)12/h2H,3H2,1H3,(H,13,14)(H2,7,8,9)(H2,10,11,12)
Formula Weight 299.0072
Chemical Name or Material (4-Amino-2-methylpyrimidin-5-yl)methyl Trihydrogen Diphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.