missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 5305-59-9 |
| Melting Point | 212.0°C to 216.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass Bottle |
| Molecular Formula | C4H4ClN3 |
| Quantity | 1 g |
| Synonym | 4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630 |
| InChI Key | DUKKRSPKJMHASP-UHFFFAOYSA-N |
| SMILES | C1=C(N=CN=C1Cl)N |
| Show More |
Safety and Handling
Product Identifier
- 4-Amino-6-chloropyrimidine, 95%
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?