missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine N-Carbamic Acid Methyl Ester, TRC

Product Code. 30325789
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325789 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325789 Supplier Toronto Research Chemicals Supplier No. TRCA583175100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminoantipyrine N-Carbamic Acid Methyl Ester
CAS 10077-96-0
Molecular Formula C13H15N3O3
Quantity 100 mg
Synonym (2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)carbamic Acid Methyl Ester, 2,3-Dimethyl-5-oxo-1-phenyl-3-pyrazoline-4-carbamic Acid Methyl Ester
InChI Formula InChI=1S/C13H15N3O3/c1-9-11(14-13(18)19-3)12(17)16(15(9)2)10-7-5-4-6-8-10/h4-8H,1-3H3,(H,14,18)
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)NC(=O)OC
IUPAC Name methyl N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)carbamate
Molecular Weight (g/mol) 261.28
Formula Weight 261.1113
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.