missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzyl Alcohol, TRC

Product Code. 30009115
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009115 5 g 5g
30006970 1 g 1g
30004762 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009115 Supplier Toronto Research Chemicals Supplier No. TRCA5871055G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-04-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
Synonym Tetracaine Impurity 3
IUPAC Name (4-aminophenyl)methanol
SMILES Nc1ccc(CO)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H9NO/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5,8H2
Formula Weight 123.0684
Chemical Name or Material 4-Aminobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.