missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobiphenyl, 98%

Product Code. 15481658
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481658 25 g 25g
15461658 1 g 1g
15471658 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15481658 Supplier Thermo Scientific Alfa Aesar Supplier No. H55424.14
  • ONLINE EXCLUSIVE PRICE
    236.00€
    212.65€ / 25g
    Save 23.35€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 92-67-1 | C12H11N | 169.227 g/mol

It is used to manufacture azo dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to manufacture azo dyes.

Solubility
Soluble in Dichloromethane, DMSO and Methanol

Notes
Store at -20°C. Avoid heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-67-1
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00007879
InChI Key DMVOXQPQNTYEKQ-UHFFFAOYSA-N
Synonym 4-aminobiphenyl, 4-aminodiphenyl, 4-biphenylamine, biphenyl-4-ylamine, 1,1'-biphenyl-4-amine, biphenyl-4-amine, xenylamine, 4-biphenylylamine, p-phenylaniline, p-aminobiphenyl
PubChem CID 7102
ChEBI CHEBI:1784
IUPAC Name 4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N

Specifications

Melting Point 52°C to 54°C
Boiling Point 191°C (15 mmHg)
Quantity 25 g
UN Number UN3077
Beilstein 386533
Merck Index 14,1235
Solubility Information Soluble in Dichloromethane,DMSO and Methanol
Formula Weight 169.23
Percent Purity 98%
Chemical Name or Material 4-Aminobiphenyl
compliance-icons compliance-icons
Product Identifier
  • 4-Aminobiphenyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.