missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Aminomethyl)-2-methoxyphenol Hydrochloride (Vanillylamine Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-(Aminomethyl)-2-methoxyphenol Hydrochloride (Vanillylamine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 4-(Aminomethyl)-2-methoxyphenol Hydrochloride (Vanillylamine Hydrochloride) |
| CAS | 10-2-7149 |
| Analyte or Component Names | 4-(Aminomethyl)-2-methoxyphenol Hydrochloride (Vanillylamine Hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H11 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(Aminomethyl)-2-methoxyphenol Hydrochloride, Vanillylamine Hydrochloride |
| InChI Formula | InChI=1S/C8H11NO2.ClH/c1-11-8-4-6(5-9)2-3-7(8)10;/h2-4,10H,5,9H2,1H3;1H |
| SMILES | Cl.COc1cc(CN)ccc1O |
| IUPAC Name | 4-(aminomethyl)-2-methoxyphenol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction