missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 95%

Product Code. 15490026
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490026 1 g 1g
15400036 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490026 Supplier Thermo Scientific Alfa Aesar Supplier No. H52691.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 850568-55-7 | C13H21BClNO2 | 269.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-55-7
Molecular Formula C13H21BClNO2
Molecular Weight (g/mol) 269.576
MDL Number MFCD02179455
InChI Key KPECMJIHZZWTJN-UHFFFAOYSA-N
Synonym 4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl
PubChem CID 16427088
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine;hydrochloride
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl

Specifications

Melting Point 228°C to 238°C
Quantity 1 g
Formula Weight 269.58
Percent Purity 95%
Chemical Name or Material 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.