missing translation for 'onlineSavingsMsg'
Learn More

4-Aminotetrahydropyran, 97%, Maybridge

Product Code. 10798245
Click to view available options
:
Amber Glass Bottle; 250mg
This item is not returnable. View return policy

Product Code. 10798245

Brand: Thermo Scientific Maybridge CC29914CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 38041-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H12NO
MDL Number MFCD02179436
Quantity 250 mg
Synonym 4-aminotetrahydropyran, tetrahydro-2h-pyran-4-amine, tetrahydro-pyran-4-ylamine, 4-aminotetrahyropyran, 4-aminotetrahydro-2h-pyran, 2h-pyran-4-amine, tetrahydro, tetrahydro-2h-pyran-4-ylamine, tetrahydropyran-4-ylamine, tetrahydropyran-4-amine, 2h-3,4,5,6-tetrahydropyran-4-ylamine
InChI Key AHVQYHFYQWKUKB-UHFFFAOYSA-O
SMILES [NH3+]C1CCOCC1
IUPAC Name oxan-4-aminium
Molecular Weight (g/mol) 102.16
PubChem CID 419223
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 4-Aminotetrahydropyran
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.