missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzoic acid, 97%, ACROS Organics™

Product Code. 11925141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925141 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11925141 Supplier Thermo Scientific Acros Supplier No. 442480050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1486-51-7
Melting Point 187.0°C to 190.0°C
Boiling Point 125.0°C (0.1 Torr)
Assay Percent Range 97%
Molecular Formula C14H12O3
MDL Number MFCD00016527
Quantity 5 g
Synonym 4-benzyloxybenzoic acid, 4-benzyloxy benzoic acid, benzoic acid, 4-phenylmethoxy, p-benzyloxy benzoic acid, 4-phenylmethoxy benzoic acid, 4-benzyloxy-benzoic acid, 4-benzyl-oxy benzoic acid, benzoic acid,4-phenylmethoxy, asischem a95731
InChI Key AQSCHALQLXXKKC-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
IUPAC Name 4-(benzyloxy)benzoic acid
Molecular Weight (g/mol) 228.25
PubChem CID 73880
Formula Weight 228.25
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Benzyloxy)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.