missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenyl 4-Hydroxyphenyl Sulfone, TRC

Product Code. 17953125
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953125 50 mg 50mg
17931215 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953125 Supplier Toronto Research Chemicals Supplier No. TRCB28784550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63134-33-8
Molecular Formula C19 H16 O4 S
Molecular Weight (g/mol) 340.39
Synonym 4-[[4-(Phenylmethoxy)phenyl]sulfonyl]phenol, 4-Benzyloxy-4'-hydroxydiphenyl sulfone, 4-Benzyloxyphenyl 4-hydroxyphenyl sulfone, 4-Benzyloxyphenyl-4'-hydroxyphenyl sulfone, 4-Hydroxy-4'-benzyloxydiphenyl sulfone, BPS-BN, BPS-MA 3, p-(p-Benzyloxyphenylsulfonyl)phenol
IUPAC Name 4-(4-phenylmethoxyphenyl)sulfonylphenol
SMILES Oc1ccc(cc1)S(=O)(=O)c2ccc(OCc3ccccc3)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H16O4S/c20-16-6-10-18(11-7-16)24(21,22)19-12-8-17(9-13-19)23-14-15-4-2-1-3-5-15/h1-13,20H,14H2
Formula Weight 340.0769
Chemical Name or Material Bisphenol S-monobenzyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.