Learn More
4-Benzylpiperidine, 98%
Product Code. 10743973 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
Product Code. Quantity Packaging unitSize
10743973 5 g Glass bottle 5g
10215880 100 g Glass bottle 100g
2 options
Product Code. 10743973
Brand: Thermo Scientific Acros 161930050
Request Bulk or Custom Format
CAS: 31252-42-3 | C12H17N | 175.28 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C(C1CCNCC1)C1=CC=CC=C1
Specifications
CAS Min % 97.5
CAS Max % 100.0
Melting Point 6.0°C to 7.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 279.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 5 g
Beilstein 20, 296
Specific Gravity 0.99
Formula Weight 175.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 4-Benzylpiperidine, 98%