Learn More
4-(Boc-aminomethyl)-N-Fmoc-L-phenylalanine, 95%
Product Code. 15469495
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469495 250 mg 250mg
15428497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15469495 Supplier Thermo Scientific Alfa Aesar Supplier No. H52008.MD
 Restricted Item
In Stock
Add to basket
CAS: 204715-91-3 | C30H32N2O6 | 516.594 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 204715-91-3
Molecular Formula C30H32N2O6
Molecular Weight (g/mol) 516.594
MDL Number MFCD01317015
InChI Key QSPDPKLGXGMDAY-SANMLTNESA-N
Synonym fmoc-4-boc-aminomethyl-l-phenylalanine, fmoc-l-4-aminomethylphe boc, 2s-3-4-tert-butoxy carbonyl amino methyl phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxycarbonyl amino methyl phenyl propanoic acid, 2s-3-4-tert-butoxycarbonyl amino methyl phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, 2r-3-4-tert-butoxycarbonyl amino methyl phenyl-2-9h-fluoren-9-yl methoxycarbonyl amino propanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-2-methylpropan-2-yl oxycarbonylamino methyl phenyl propanoic acid, s-3-4-tert-butoxycarbonylamino-methyl-phenyl-2-9h-fluoren-9-ylmethoxycarbonylamino-propionic acid, 4-1,1-dimethylethoxy carbonyl amino methyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalanine, l-phenylalanine,4-1,1-dimethylethoxy carbonyl amino methyl-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 2761481
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Specifications
Quantity 250 mg
Formula Weight 516.59
Percent Purity 95%
Chemical Name or Material 4-(Boc-aminomethyl)-N-Fmoc-L-phenylalanine