Learn More
4-Bromo-1-methoxy-2-nitrobenzene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 33696-00-3 |
| Molecular Formula | C7H6BrNO3 |
| MDL Number | MFCD00055529 |
| Quantity | 500 g |
| SMILES | COC1=CC=C(Br)C=C1[N+]([O-])=O |
| Molecular Weight (g/mol) | 232.03 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Bromo-1-methoxy-2-nitrobenzene |