Learn More
4-Bromo-2-picoline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-methylpyridine |
| CAS | 22282-99-1 |
| Molecular Formula | C6 H6 Br N |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H6BrN/c1-5-4-6(7)2-3-8-5/h2-4H,1H3 |
| SMILES | Cc1cc(Br)ccn1 |
| IUPAC Name | 4-bromo-2-methylpyridine |
| Molecular Weight (g/mol) | 172.02 |
| Formula Weight | 170.9684 |
Product Title
Haben Sie Verbesserungsvorschläge?